Kaliumthioacetat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Potassium ethanethioate
L05405.14
L05405.22
L05405.14EA
57.8
EUR
L05405.14
L05405.22
Kaliumthioacetat ≥98%
Kaliumthioacetat
Purity: 98%
CAS No.: 10387-40-3
Molecular Formula: C2H3KOS
Formula Weight: 114.21
Odour: Stench
Melting Point: ca 240° dec.
Beilstein Reference: 3595448
UN No.: 3335
MDL No.: MFCD00083065
Formel:
C₂H₃KOS Molecular Weight: 114,21 g/mol Schmelzpunkt: ∼ 240 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00137704 CAS-Nummer: 10387-40-3 EINECS: 233-848-7 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...