3-(4-Fluorbenzoyl)propionsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

A13755.06 A13755.14
A13755.06EA 26.2 EUR
A13755.06 A13755.14
3-(4-Fluorbenzoyl)propionsäure ≥97%
3-(4-Fluorbenzoyl)propionsäure

Purity: 97% CAS No.: 366-77-8 Molecular Formula: C10H9FO3 Formula Weight: 196.18 Odour: Odourless Melting Point: 99-103° Beilstein Reference: 958145 MDL No.: MFCD00002793

Formel: C₁₀H₉FO₃
Schmelzpunkt: 99…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002793
CAS-Nummer: 366-77-8
EINECS: 206-679-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR