3-(4-Fluorbenzoyl)propionsäure

Lieferant: CHEMPUR

FL3846.10 FL3846.25
CHMPFL3846.10EA 0 EUR
CHMPFL3846.10 CHMPFL3846.25
3-(4-Fluorbenzoyl)propionsäure
3-(4-Fluorbenzoyl)propionsäure
Formel: C₁₀H₉FO₃
MDL: MFCD00002793
CAS-Nummer: 366-77-8
EINECS: 206-679-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR