Dimethoxymethylvinylsilan ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Dimethoxy(methyl)(vinyl)silane
L14035.22
L14035.14
AB173857.0100
L14035.22EA
204
EUR
L14035.22
L14035.14
ABCRAB173857.0100
Dimethoxymethylvinylsilan ≥97%
Dimethoxymethylvinylsilan
Purity: 97%
CAS No.: 16753-62-1
Molecular Formula: C5H12O2Si
Formula Weight: 132.23
Flash Point: 5°(41°F)
Refractive Index: 1.3950
Boiling Point: 102-104°
Beilstein Reference: 3043765
UN No.: 1993
MDL No.: MFCD00129170
Formel:
C₅H₁₂O₂Si Molecular Weight: 132,23 g/mol Siedepunkt: 102…104 °C Dichte: 0,889 Flammpunkt: 5 °C (41 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00129170 CAS-Nummer: 16753-62-1 EINECS: 240-816-6 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...