Dimethoxymethylvinylsilan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dimethoxy(methyl)(vinyl)silane

M-Clarity™ Qualitätsstufe = MQ100

446203-100ML 446203-25ML
SIAL446203-100MLEA 411 EUR
SIAL446203-100ML SIAL446203-25ML
Dimethoxymethylvinylsilan, Sigma-Aldrich®
Dimethoxymethylvinylsilan
Formel: C₅H₁₂O₂Si
Molecular Weight: 132,23 g/mol
MDL: MFCD00129170
CAS-Nummer: 16753-62-1
EINECS: 240-816-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR