Dimethoxymethylvinylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Dimethoxy(methyl)(vinyl)silane

SILS21850.10 SILS21850.25 SILS21850.100
SILNSILS21850.10EA 0 EUR
SILNSILS21850.10 SILNSILS21850.25 SILNSILS21850.100
Dimethoxymethylvinylsilan
Dimethoxymethylvinylsilan
Formel: C₅H₁₂O₂Si
Molecular Weight: 132,23 g/mol
MDL: MFCD00129170
CAS-Nummer: 16753-62-1
EINECS: 240-816-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR