9-Acridon 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 9,10-Dihydro-9-oxoacridin, 9(10H)-Acridanone, Acridone, 9(10H)-Acridone, Acridin-9(10H)-one, 9-Acridone

400250100 400250250
ACRO400250100EA 119 EUR
ACRO400250100 ACRO400250250
9-Acridon 99%
9-Acridon
Formel: C₁₃H₉NO
Molecular Weight: 195,22 g/mol
Schmelzpunkt: 360…362 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005019
CAS-Nummer: 578-95-0
EINECS: 209-434-7

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Titration with TBAH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR