9-Acridon 99
Lieferant: Thermo Fisher Scientific
Synonyme:
9,10-Dihydro-9-oxoacridin, 9(10H)-Acridanone, Acridone, 9(10H)-Acridone, Acridin-9(10H)-one, 9-Acridone
A15222.06
A15222.14
A15222.06EA
58
EUR
A15222.06
A15222.14
9-Acridon 99
9-Acridon
Purity: 99%
CAS No.: 578-95-0
Molecular Formula: C13H9NO
Formula Weight: 195.22
Melting Point: 360-362°
Beilstein Reference: 7104
Formel:
C₁₃H₉NO Molecular Weight: 195.22 g/mol Schmelzpunkt: 360…362 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005019 CAS-Nummer: 578-95-0 EINECS: 209-434-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...