9-Acridon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 9,10-Dihydro-9-oxoacridin, 9(10H)-Acridanone, Acridone, 9(10H)-Acridone, Acridin-9(10H)-one, 9-Acridone

M-Clarity™ Qualitätsstufe = MQ100

150215-1G 150215-5G
SIAL150215-1GEA 37.8 EUR
SIAL150215-1G SIAL150215-5G
9-Acridon, Sigma-Aldrich®
9-Acridon
Formel: C₁₃H₉NO
Molecular Weight: 195,22 g/mol
MDL: MFCD00005019
CAS-Nummer: 578-95-0
EINECS: 209-434-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR