1-Aza-15-krone-5 (1,4,7,10-Tetraoxa-13-azacyclopentadecan) ≥97%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: 1,4,7,10-Tetraoxa-13-azacyclopentadecane, 1-Aza-15-crown 5-Ether, 1,4,7,10-Tetraoxa-13-azacyclopentadecane
H51064.MD H51064.03 H51064.06
H51064.MDEA 73 EUR
H51064.MD H51064.03 H51064.06
1-Aza-15-krone-5 (1,4,7,10-Tetraoxa-13-azacyclopentadecan) ≥97%
1-Aza-15-krone-5 (1,4,7,10-Tetraoxa-13-azacyclopentadecan)
Purity: 97% CAS No.: 66943-05-3 Molecular Formula: C10H21NO4 Formula Weight: 219.28 Beilstein Reference: 971959 MDL No.: MFCD00075465
Formel: C₁₀H₂₁NO₄
Molecular Weight: 219.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075465
CAS-Nummer: 66943-05-3
EINECS: 266-523-3