Benzo-15-krone-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecan)

Lieferant: FLUOROCHEM

002501-1G 002501-5G
FLUO002501-1GEA 75.6 EUR
FLUO002501-1G FLUO002501-5G
Benzo-15-krone-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecan)
Benzo-15-krone-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecan)
Formel: C₁₄H₂₀O₅
MDL: MFCD00005945
CAS-Nummer: 14098-44-3
EINECS: 237-947-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR