1,10-Diaza-18-krone-6 (1,4,10,13-Tetraoxa-7,16-diazacyclooctadecan, Kryptofix® 22) 96%
Lieferant: Thermo Scientific
Achtung
Synonyme: Diaza-18-crown-6, 1,4,10,13-Tetraoxa-7,16-diazacyclooctadecane
H51063.MD H51063.03 H51063.06
H51063.MDEA 56 EUR
H51063.MD H51063.03 H51063.06
1,10-Diaza-18-krone-6 (1,4,10,13-Tetraoxa-7,16-diazacyclooctadecan, Kryptofix® 22) 96%
1,10-Diaza-18-krone-6 (1,4,10,13-Tetraoxa-7,16-diazacyclooctadecan, Kryptofix® 22)
Purity: 96% CAS No.: 23978-55-4 Molecular Formula: C12H26N2O4 Formula Weight: 262.35 Beilstein Reference: 609764 MDL No.: MFCD00005112
Formel: C₁₂H₂₆N₂O₄
Molecular Weight: 262,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005112
CAS-Nummer: 23978-55-4
EINECS: 245-965-0