Dibenzo-24-krone-8 (Dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosan), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dibenzo-24-crown-8

M-Clarity™ Qualitätsstufe = MQ100

253197-1G 253197-5G
SIAL253197-1GEA 88.8 EUR
SIAL253197-1G SIAL253197-5G
Dibenzo-24-krone-8 (Dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosan), Sigma-Aldrich®
Dibenzo-24-krone-8 (Dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosan)
Formel: C₂₄H₃₂O₈
Molecular Weight: 448.51 g/mol
MDL: MFCD00005101
CAS-Nummer: 14174-09-5
EINECS: 238-027-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR