(3-Glycidoxypropyl)triethoxysilan

Lieferant: ABCR

Synonyme: (3-Glycidyloxypropyl)triethoxysilan, Geniosil GF 82, (3-(2,3-Epoxypropoxy)propyl)triethoxysilan

SIG5829.0 AB153338.2000
ABCRSIG5829.0EA 0 EUR
ABCRSIG5829.0 ABCRAB153338.2000
(3-Glycidoxypropyl)triethoxysilan
(3-Glycidoxypropyl)triethoxysilan
Formel: C₁₂H₂₆O₅Si
Molecular Weight: 278,42 g/mol
MDL: MFCD01311705
CAS-Nummer: 2602-34-8
EINECS: 220-011-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR