2-Chlor-1,1,2-trifluor-1-methoxyethan
Lieferant: FLUOROCHEM
004532-5G
004532-25G
FLUO004532-5GEA
31.1
EUR
FLUO004532-5G
FLUO004532-25G
2-Chlor-1,1,2-trifluor-1-methoxyethan
2-Chlor-1,1,2-trifluor-1-methoxyethan
Formel:
C₃H₄ClF₃O |
MDL:
MFCD00039335 CAS-Nummer: 425-87-6 EINECS: 207-038-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...