1,1,3,3-Tetramethyldisiloxan
Lieferant: ABCR
AB111418.0250
AB111418.0025
ABCRAB111418.0250EA
0
EUR
ABCRAB111418.0250
ABCRAB111418.0025
1,1,3,3-Tetramethyldisiloxan
1,1,3,3-Tetramethyldisiloxan
Formel:
C₄H₁₄OSi₂ Molecular Weight: 134,33 g/mol Siedepunkt: 71 °C (1013 hPa) Dichte: 0,7555 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00008256 CAS-Nummer: 3277-26-7 EINECS: 221-906-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...