Eriochromschwarz T 1% in Natriumchlorid

Lieferant: Bernd Kraft
Achtung

Synonyme: 1-(2-Hydroxy-1-naphthylazo)-6-nitro-2-naphthol-4-sulfonsäure Natriumsalz, Natrium-3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalinsulfonat

04124.0600 04124.0602 04124.0601
KRAF04124.0600EA 79.4 EUR
KRAF04124.0600 KRAF04124.0602 KRAF04124.0601
Eriochromschwarz T 1% in Natriumchlorid
Eriochromschwarz T
Formel: C₂₀H₁₂N₃NaO₇S
Molecular Weight: 461,39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003935
CAS-Nummer: 1787-61-7
EINECS: 217-250-3
UN: 3077
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR