α'-Chlor-α,α,α-trifluor-p-xylol
Lieferant: CHEMPUR
Synonyme:
4-(Trifluormethyl)benzylchlorid, 4-(Chlormethyl)benzotrifluorid, 1-(Chlormethyl)-4-(trifluormethyl)benzol
SPB01488.250
SPB01488.1
CHMPSPB01488.250EA
0
EUR
CHMPSPB01488.250
MAYBSPB01488.1
α'-Chlor-α,α,α-trifluor-p-xylol
α'-Chlor-α,α,α-trifluor-p-xylol
Formel:
C₈H₆ClF₃ Molecular Weight: 194,58 g/mol Siedepunkt: 47…49 °C (20 mmHg) Schmelzpunkt: 19 °C Dichte: 1,31 g/cm³ (20 °C) |
MDL:
MFCD00040772 CAS-Nummer: 939-99-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...