3-Amino-2-chlor-4-methylpyridin
Lieferant: FLUOROCHEM
Synonyme:
3-Amino-2-chloro-4-methylpyridine, 3-Amino-2-chloro-4-picoline, 2-Chloro-4-methylpyridin-3-amine
021733-5G
021733-25G
FLUO021733-5GEA
78.6
EUR
FLUO021733-5G
FLUO021733-25G
3-Amino-2-chlor-4-methylpyridin
3-Amino-2-chloro-4-methylpyridine
Formel:
C₆H₇ClN₂ Molecular Weight: 142,59 g/mol |
MDL:
MFCD00673152 CAS-Nummer: 133627-45-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...