Hydroxylamin-Lösung 50% (w/w) in Wasser 99.999%, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

SIAL467804-10MLEA 35.8 EUR
SIAL467804-10ML SIAL467804-50ML
Hydroxylamin-Lösung 50% (w/w) in Wasser 99.999%, Sigma-Aldrich®
Hydroxylamin

Hydroxylamine is a weak base that is mainly used as a reducing agent in organic reactions.


Reactant for preparation of:
Prodrug for cardiovascular agent Nω-hydroxy-L-arginine (NOHA, nitric oxide precursor)
Hydroxyaminoguanidines as anti-cancer agents
Nonsteroidal 2,3-dihydroquinoline glucocorticoid receptor agonists with reduced phosphoenolpyruvate caboxykinase (PEPCK) transactivation
Carboxamide derivatives of ofloxacin with improved antimicrobial properties
Analogues of coumarin based TNF-α converting enzyme (TACE) inhibitors
HIV integrase inhibitors

Formel: H₃NO
Molecular Weight: 33,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00044522
CAS-Nummer: 7803-49-8
EINECS: 232-259-2
UN: 3266
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR