Oxalsäureethylesterchlorid ≥98%

Lieferant: ABCR
Gefahr

Synonyme: Ethyl oxalyl monochloride, Oxalic Acid Monoethyl Ester Chloride, Ethyl Chlorooxoacetate, Ethyl Chloroglyoxylate, Chloroglyoxylic Acid Ethyl Ester, Ethyl chloroformylformate

14653-0500 AB108675 A14653.22 A14653.14 A14653.36 A14653.0B
ABCR14653-0500EA 0 EUR
ABCR14653-0500 ABCRAB108675 A14653.22 A14653.14 A14653.36 A14653.0B
Oxalsäureethylesterchlorid ≥98%
Oxalsäureethylesterchlorid

Purity: 98% CAS No.: 4755-77-5 Molecular Formula: C4ClH5O3 Formula Weight: 136.53 Flash Point: 41°(105°F) Refractive Index: 1.4160 Boiling Point: 134-135° Beilstein Reference: 506725 UN No.: 2920 MDL No.: MFCD00000706

Formel: C₄H₅ClO₃
Molecular Weight: 136,53 g/mol
Siedepunkt: 134…135 °C
Dichte: 1,226
Flammpunkt: 41 °C (105 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000706
CAS-Nummer: 4755-77-5
EINECS: 225-285-0
UN: 2920
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR