Bromhydrochinon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Bromobenzene-1,4-diol
167134-10G
167134-1G
SIAL167134-10GEA
211
EUR
SIAL167134-10G
SIAL167134-1G
Bromhydrochinon, Sigma-Aldrich®
Bromhydrochinon
Formel:
C₆H₅BrO₂ Molecular Weight: 189,01 g/mol |
MDL:
MFCD00041747 CAS-Nummer: 583-69-7 EINECS: 209-516-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...