2,3-Dimethylhydrochinon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,4-Dihydroxy-2,3-dimethylbenzene
M-Clarity™ Qualitätsstufe = MQ100
300756-25G
300756-5G
SIAL300756-25GEA
581
EUR
SIAL300756-25G
SIAL300756-5G
2,3-Dimethylhydrochinon, Sigma-Aldrich®
2,3-Dimethylhydrochinon
Formel:
C₈H₁₀O₂ Molecular Weight: 138.17 g/mol |
MDL:
MFCD00009997 CAS-Nummer: 608-43-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...