2-Chlor-4-ethylpyrimidin

Lieferant: COMBI-BLOCKS

HC-6168-5G HC-6168-1G
COBBHC-6168-5GEA 796 EUR
COBBHC-6168-5G COBBHC-6168-1G
2-Chlor-4-ethylpyrimidin
2-Chloro-4-ethylpyrimidine
Formel: C₆H₇ClN₂
MDL: MFCD10699651
CAS-Nummer: 188707-99-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR