4,6-Dichlor-2-methylpyrimidin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Methyl-4,6-dichloropyrimidine
M-Clarity™ Qualitätsstufe = MQ100
596728-25G
596728-5G
SIAL596728-25GEA
83.4
EUR
SIAL596728-25G
SIAL596728-5G
4,6-Dichlor-2-methylpyrimidin, Sigma-Aldrich®
4,6-Dichloro-2-methylpyrimidine
Formel:
C₅H₄Cl₂N₂ Molecular Weight: 163,01 g/mol |
MDL:
MFCD00090472 CAS-Nummer: 1780-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...