2-Chlor-5-fluorpyrimidin

Lieferant: COMBI-BLOCKS

OS-7507-25G OS-7507-1G OS-7507-5G
COBBOS-7507-25GEA 193 EUR
COBBOS-7507-25G COBBOS-7507-1G COBBOS-7507-5G
2-Chlor-5-fluorpyrimidin
2-Chloro-5-fluoropyrimidine
Formel: C₄H₂ClFN₂
Molecular Weight: 132,52 g/mol
MDL: MFCD03788197
CAS-Nummer: 62802-42-0
UN: 3267
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR