Aluminiumtri-tert-butoxid ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Aluminium tert-butoxide, Aluminumtert-butoxide

89891.06 89891.14 89891.22
89891.06EA 54.2 EUR
89891.06 89891.14 89891.22
Aluminiumtri-tert-butoxid ≥97%
Aluminiumtri-tert-butoxid

Purity: 97% CAS No.: 556-91-2 Molecular Formula: Al[OC(CH3)3]3 Formula Weight: 246.32 Physical Form: Powder Density: 1.0251 Melting Point: 200-202° Boiling Point: 156°/2mm Solubility: Very soluble in organic solvents Beilstein Reference: 4148229 Merck Reference: 14,333 UN No.: 2925 MDL No.: MFCD00008801

Formel: C₁₂H₂₇AlO₃
Molecular Weight: 246,33 g/mol
Siedepunkt: 156 °C (2 mmHg)
Schmelzpunkt: 200…202 °C
Dichte: 1,025
Lagertemperatur: Raumtemperatur
MDL: MFCD00008801
CAS-Nummer: 556-91-2
EINECS: 209-146-1
ADR: 4.1,III
Merck Index: 14,333

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR