3-Methyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

Synonyme: 3-Methylpyrazole

OS-7608-5G OS-7608-25G OS-7608-100G
COBBOS-7608-5GEA 25.5 EUR
COBBOS-7608-5G COBBOS-7608-25G COBBOS-7608-100G
3-Methyl-1H-pyrazol
3-Methyl-1H-pyrazole
Formel: C₄H₆N₂
Molecular Weight: 82,11 g/mol
MDL: MFCD00005240
CAS-Nummer: 1453-58-3
EINECS: 215-925-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR