3-Methyl-1H-pyrazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Methylpyrazole
M-Clarity™ Qualitätsstufe = MQ100
M75802-25G
M75802-5G
SIALM75802-25GEA
58.2
EUR
SIALM75802-25G
SIALM75802-5G
3-Methyl-1H-pyrazol, Sigma-Aldrich®
3-Methyl-1H-pyrazole
Formel:
C₄H₆N₂ Molecular Weight: 82,11 g/mol |
MDL:
MFCD00005240 CAS-Nummer: 1453-58-3 EINECS: 215-925-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...