3-Phenyl-1H-pyrazol

Lieferant: FLUOROCHEM

Synonyme: 3(5)-Phenyl-1H-pyrazole, 3-Phenylpyrazole

CFF675.5 017112-25G 017112-1G 017112-5G
CHMPCFF675.5EA 587 EUR
CHMPCFF675.5 FLUO017112-25G FLUO017112-1G FLUO017112-5G
3-Phenyl-1H-pyrazol
3-Phenyl-1H-pyrazole
Formel: C₉H₈N₂
Molecular Weight: 144,18 g/mol
MDL: MFCD00159654
CAS-Nummer: 2458-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR