2-Piperidinmethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(Hydroxymethyl)piperidine

M-Clarity™ Qualitätsstufe = MQ100

155225-5G 155225-100G 155225-25G
SIAL155225-5GEA 77.6 EUR
SIAL155225-5G SIAL155225-100G SIAL155225-25G
2-Piperidinmethanol, Sigma-Aldrich®
2-Piperidinmethanol
Formel: C₆H₁₃NO
Molecular Weight: 115,18 g/mol
MDL: MFCD00005987
CAS-Nummer: 3433-37-2
EINECS: 222-333-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR