3-Piperidinmethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(Hydroxymethyl)piperidine

M-Clarity™ Qualitätsstufe = MQ200

155233-25G 155233-5G
SIAL155233-25GEA 76.8 EUR
SIAL155233-25G SIAL155233-5G
3-Piperidinmethanol, Sigma-Aldrich®
3-Piperidinmethanol
Formel: C₆H₁₃NO
Molecular Weight: 115,18 g/mol
MDL: MFCD00005997
CAS-Nummer: 4606-65-9
EINECS: 225-013-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR