4-Piperidinmethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Piperidin-4-ylmethanol

M-Clarity™ Qualitätsstufe = MQ100

497312-25G 497312-5G
SIAL497312-25GEA 696 EUR
SIAL497312-25G SIAL497312-5G
4-Piperidinmethanol, Sigma-Aldrich®
4-Piperidinmethanol
Formel: C₆H₁₃NO
Molecular Weight: 115,18 g/mol
MDL: MFCD00174228
CAS-Nummer: 6457-49-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR