Ethylbutyrylacetat ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Ethyl Butyrylacetate, Ethyl 3-oxohexanoate

A12786.18 A12786.30
A12786.18EA 35 EUR
A12786.18 A12786.30
Ethylbutyrylacetat ≥98%
Ethylbutyrylacetat

Purity: 98% CAS No.: 3249-68-1 Molecular Formula: C8H14O3 Formula Weight: 158.20 Flash Point: 78°(172°F) Refractive Index: 1.4270 Melting Point: -44° Boiling Point: 94-95°/12mm Beilstein Reference: 507689 MDL No.: MFCD00009401

Formel: C₈H₁₄O₃
Molecular Weight: 158,2 g/mol
Siedepunkt: 94…95 °C (12 mmHg)
Schmelzpunkt: –44 °C
Dichte: 0,990
Flammpunkt: 78 °C (172 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009401
CAS-Nummer: 3249-68-1
EINECS: 221-835-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR