Ethylbutyrylacetat
Lieferant: Thermo Scientific
Synonyme:
Ethyl Butyrylacetate, Ethyl 3-oxohexanoate
SB00666.1
SB00666.10
SB00666.50
MAYBSB00666.1EA
0
EUR
MAYBSB00666.1
MAYBSB00666.10
MAYBSB00666.50
Ethylbutyrylacetat
Ethylbutyrylacetat
Formel:
C₈H₁₄O₃ Molecular Weight: 158.2 g/mol |
MDL:
MFCD00009401 CAS-Nummer: 3249-68-1 EINECS: 221-835-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...