Dihydrocumarin ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Chroman-2-One, 1,2-Benzodihydropyrone
B21229.0E
B21229.22
B21229.36
B21229.0EEA
384
EUR
B21229.0E
B21229.22
B21229.36
Dihydrocumarin ≥99%
Dihydrocumarin
Purity: 99%
CAS No.: 119-84-6
Molecular Formula: C9H8O2
Formula Weight: 148.16
Flash Point: 130°(266°F)
Refractive Index: 1.5560
Melting Point: 24-25°
Boiling Point: 272°
Beilstein Reference: 4584
MDL No.: MFCD00006881
Formel:
C₉H₈O₂ Molecular Weight: 148,16 g/mol Siedepunkt: 272 °C Schmelzpunkt: 24…25 °C Dichte: 1,169 Flammpunkt: 130 °C (266 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006881 CAS-Nummer: 119-84-6 EINECS: 204-354-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...