Dihydrocumarin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Chroman-2-One, 1,2-Benzodihydropyrone

M-Clarity™ Qualitätsstufe = MQ100

D104809-100G D104809-5G
SIALD104809-100GEA 58.8 EUR
SIALD104809-100G SIALD104809-5G
Dihydrocumarin, Sigma-Aldrich®
Dihydrocumarin
Formel: C₉H₈O₂
Molecular Weight: 148,16 g/mol
MDL: MFCD00006881
CAS-Nummer: 119-84-6
EINECS: 204-354-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR