Natrium-tert-butanolat ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Sodium tert-butoxide, Sodium t-butoxide, Sodium Tert-Butylate
A19942.22
A19942.0E
A19942.36
A19942.22EA
53.2
EUR
A19942.22
A19942.0E
A19942.36
Natrium-tert-butanolat ≥97%
Natrium-tert-butanolat
Purity: 97%
CAS No.: 865-48-5
Molecular Formula: C4H9NaO
Formula Weight: 96.11
Odour: Odourless
Beilstein Reference: 3654215
UN No.: 3206
MDL No.: MFCD00040575
Formel:
C₄H₉NaO Molecular Weight: 96.1 g/mol Dichte: 0.785 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040575 CAS-Nummer: 865-48-5 EINECS: 212-741-9 UN: 3206 ADR: 4.2 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...