Natrium-tert-butanolat, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Sodium tert-butoxide, Sodium t-butoxide, Sodium Tert-Butylate
703788-25G
703788-5G
SIAL703788-25GEA
160
EUR
SIAL703788-25G
SIAL703788-5G
Natrium-tert-butanolat, Sigma-Aldrich®
Natrium-tert-butanolat
Formel:
C₄H₉NaO Molecular Weight: 96.1 g/mol |
MDL:
MFCD00040575 CAS-Nummer: 865-48-5 EINECS: 212-741-9 UN: 3206 ADR: 4.2 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...