D(-)-Ornithin Hydrochlorid

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-D-Orn-OH·HCl, H-D-Orn-OH.HCl

4031134.0005 4031134.0025 4031134.0100
BACE4031134.0005EA 0 EUR
BACE4031134.0005 BACE4031134.0025 BACE4031134.0100
D(-)-Ornithin Hydrochlorid
D(-)-Ornithin Hydrochlorid
Formel: C₅H₁₂N₂O₂·HCl
Molecular Weight: 168.62 g/mol
MDL: MFCD00012917
CAS-Nummer: 16682-12-5
EINECS: 240-729-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR