L(+)-Ornithin Hydrochlorid

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-Orn-OH Hydrochloride, H-Orn-OH·HCl

4031133.0250 4031133.1000
BACE4031133.0250EA 0 EUR
BACE4031133.0250 BACE4031133.1000
L(+)-Ornithin Hydrochlorid
L(+)-Ornithin Hydrochlorid
Formel: C₅H₁₂N₂O₂·HCl
Molecular Weight: 168.62 g/mol
MDL: MFCD00064562
CAS-Nummer: 3184-13-2
EINECS: 221-678-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR