2-(Ethylsulfanyl)ethanol

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-hydroxyethyl sulphide, 2-(Ethylmercapto)-ethanol

F002951-100G 002951-1KG
FLUOF002951-100GEA 108 EUR
FLUOF002951-100G FLUO002951-1KG
2-(Ethylsulfanyl)ethanol
2-(Ethylthio)ethanol
Formel: C₄H₁₀OS
Molecular Weight: 106.19 g/mol
MDL: MFCD00002909
CAS-Nummer: 110-77-0
EINECS: 203-802-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR