2-(Methylthio)ethanol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

L05442.06 L05442.14 L05442.22
L05442.06EA 24 EUR
L05442.06 L05442.14 L05442.22
2-(Methylthio)ethanol ≥99%
2-(Methylthio)ethanol

Purity: 99% CAS No.: 5271-38-5 Molecular Formula: C3H8OS Formula Weight: 92.16 Flash Point: 70°(158°F) Refractive Index: 1.4944 Odour: Stench Boiling Point: 169-171° Beilstein Reference: 1731081 UN No.: 3334 MDL No.: MFCD00002908

Formel: C₃H₈OS
Siedepunkt: 169…171 °C
Dichte: 1,060
Flammpunkt: 70 °C (158 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002908
CAS-Nummer: 5271-38-5
EINECS: 226-090-3
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR