Ethylisobutyrylacetat ≥94%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Ethyl 4-methyl-3-oxopentanoate
L09016.06
L09016.14
AB171496.0025
L09016.06EA
24.5
EUR
L09016.06
L09016.14
ABCRAB171496.0025
Ethylisobutyrylacetat ≥94%
Ethylisobutyrylacetat
Purity: 94%
CAS No.: 7152-15-0
Molecular Formula: C8H14O3
Formula Weight: 158.20
Flash Point: 53°(127°F)
Refractive Index: 1.4265
Melting Point: -9°
Boiling Point: 172-174°
Beilstein Reference: 636726
UN No.: 3272
MDL No.: MFCD00009198
Formel:
C₈H₁₄O₃ Molecular Weight: 158,2 g/mol Siedepunkt: 172…174 °C Schmelzpunkt: –9 °C Dichte: 0,980 Flammpunkt: 53 °C (127 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009198 CAS-Nummer: 7152-15-0 EINECS: 230-491-9 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...