3,3-Dimethylbutyraldehyd, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3,3-Dimethylbutanal
M-Clarity™ Qualitätsstufe = MQ100
359904-1ML
359904-5ML
SIAL359904-1MLEA
60.3
EUR
SIAL359904-1ML
SIAL359904-5ML
3,3-Dimethylbutyraldehyd, Sigma-Aldrich®
3,3-Dimethylbutyraldehyd
Formel:
C₆H₁₂O |
MDL:
MFCD00042807 CAS-Nummer: 2987-16-8 EINECS: 221-054-3 UN: 1989 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...