Thiazolidin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H26427.03 H26427.06
H26427.03EA 15 EUR
H26427.03 H26427.06
Thiazolidin ≥97%
Thiazolidin

Purity: 97% CAS No.: 504-78-9 Molecular Formula: C3H7NS Formula Weight: 89.16 Flash Point: 56°(133°F) Refractive Index: 1.5545 Odour: Characteristic Boiling Point: 165° UN No.: 1993 MDL No.: MFCD00005211

Formel: C₃H₇NS
Molecular Weight: 89.16 g/mol
Siedepunkt: 165 °C
Dichte: 1.131
Flammpunkt: 56 °C (133 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005211
CAS-Nummer: 504-78-9
EINECS: 208-002-5
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR