Thiazolidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

149691-1G 149691-5G
SIAL149691-1GEA 44.7 EUR
SIAL149691-1G SIAL149691-5G
Thiazolidin, Sigma-Aldrich®
Thiazolidin
Formel: C₃H₇NS
Molecular Weight: 89.16 g/mol
MDL: MFCD00005211
CAS-Nummer: 504-78-9
EINECS: 208-002-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR