2,5-Piperazindion

Lieferant: CHEMPUR

Synonyme: Glycine anhydride, Piperazine-2,5-dione

XBX00129.250 XBX00129.1
CHMPXBX00129.250EA 0 EUR
CHMPXBX00129.250 MAYBXBX00129.1
2,5-Piperazindion
2,5-Piperazindion
Formel: C₄H₆N₂O₂
Molecular Weight: 114,1 g/mol
MDL: MFCD00006009
CAS-Nummer: 106-57-0
EINECS: 203-411-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR