4-Aminobutyraldehyddiethylacetal 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4,4-Diethoxybutan-1-amine, 4-Aminobutanal diethyl acetal
103260050
103260250
103261000
ACRO103260050EA
34.2
EUR
ACRO103260050
ACRO103260250
ACRO103261000
4-Aminobutyraldehyddiethylacetal 95%
4-Aminobutyraldehyddiethylacetal
Formel:
C₈H₁₉NO₂ Molecular Weight: 161,24 g/mol Siedepunkt: 196 °C (760 mmHg) Dichte: 0,9 g/cm³ Flammpunkt: 62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008227 CAS-Nummer: 6346-09-4 EINECS: 228-753-2 UN: 2735 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4270 to 1.4310 (20°C, 589 nm) |
GC | ≥94 % |
Solubility | (10% in H2O) Clear colorless solution |
Titration with HClO4 | 99.0 to 101.0 % (On dry substance) |
Chloride (Cl) | ≤200 ppm |
Sulfate (SO4) | ≤200 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...