4-Aminobutyraldehyddimethylacetal ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Aminobutyraldehyde Dimethyl Acetal, 4,4-Dimethoxybutylamine

B21072.06 B21072.14
B21072.06EA 50.9 EUR
B21072.06 B21072.14
4-Aminobutyraldehyddimethylacetal ≥98%
4-Aminobutyraldehyddimethylacetal

Purity: 98+% CAS No.: 19060-15-2 Molecular Formula: C6H15NO2 Formula Weight: 133.19 Flash Point: 59°(138°F) Refractive Index: 1.4300 Boiling Point: 85°/22mm UN No.: 2734 MDL No.: MFCD00189373

Formel: C₆H₁₅NO₂
Siedepunkt: 85 °C (22 mmHg)
Flammpunkt: 59 °C (138 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00189373
CAS-Nummer: 19060-15-2
EINECS: 407-690-6
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR